Would you like to log in again?
TBARS Assay Kit, Cayman Chemical
Supplier: Cayman Chemical
10009055-96
75816-718EA 314.31 USD InStock
75816-718
TBARS Assay Kit, Cayman Chemical
Assays Cellular Assays
For the measurement of lipid peroxidation in plasma, serum, urine, tissue homogenates, and cell lysates.
  • Measure TBARS in plasma, serum, urine, tissue homogenates, and cell lysates
  • Assay 40 samples in duplicate
  • Assay range: 0.0625-50 µM
  • Plate-based colorimetric measurement (530-540 nm) or fluorometric measurement (ex 530 nm, em 550 nm)
Oxidative stress in the cellular environment results in the formation of highly reactive and unstable lipid hydroperoxides. Decomposition of the unstable peroxides derived from PUFAs results in the formation of malondialdehyde (MDA), which can be quantified colorimetrically following its controlled reaction with thiobarbituric acid. The measurement of these 'Thiobarbituric Acid Reactive Substances' (TBARS) is a well-established method for screening and monitoring lipid peroxidation. Modifications of the TBARS assay has been used to evaluate a wide range of samples that include human and animal tissues and fluids, drugs and foods. Even though there remains a controversy cited in literature regarding the specificity of TBARS toward compounds other than MDA, it still remains the most widely employed assay used to determine lipid peroxidation. Cayman’s TBARS Assay Kit provides a simple, reproducible, and standardized tool for assaying lipid peroxidation in plasma, serum, urine, tissue homogenates, and cell lysates.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty