Would you like to log in again?
JC-1 Mitochondrial Membrane Potential Assay Kit, Cayman Chemical
Supplier: Cayman Chemical
10009172-1
75816-722EA 335.76 USD InStock
75816-722
JC-1 Mitochondrial Membrane Potential Assay Kit, Cayman Chemical
Assays Cellular Assays
For analysis of mitochondrial membrane potential in whole cells.
  • Measure mitochondrial membrane potential as an indicator of cell health
  • Utilizes JC-1, a fluorescent, lipophilic, cationic dye
  • Red fluorescence indicates healthy mitochondria and green fluorescence indicates mitochondria in poor health
  • Staining in less than one hour
Mitochondrial membrane potential, (ΔψM), is an important parameter of mitochondrial function used as an indicator of cell health. JC-1 is a lipophilic, cationic dye that can selectively enter into mitochondria and reversibly change color from green to red as the membrane potential increases. In healthy cells with high mitochondrial (ΔψM), JC-1 spontaneously forms complexes known as J-aggregates with intense red fluorescence. On the other hand, in apoptotic or unhealthy cells with low (ΔψM), JC-1 remains in the monomeric form, which shows only green fluorescence. Cayman’s JC-1 Mitochondrial Membrane Potential Assay Kit provides all the necessary reagents, as well as complete instructions, for analysis of mitochondrial integrity in whole cells. The assay is applicable to flow cytometry, fluorescence microscopy, and 96-well plate formats.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty