Would you like to log in again?
Leukotriene B4 Express ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical
10009292-96 10009292-480
75816-734EA 454.11 USD InStock
75816-734 75816-732
Leukotriene B4 Express ELISA Kit, Cayman Chemical Company
Assays ELISAs
A competitive assay that can be used for quantification of LTB4 in plasma and other sample matrices.
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure LTB4 levels down to 45 pg/ml
  • Rapid assay; get results in under five hours
LTB4 stimulates a number of leukocyte functions, including aggregation, stimulation of ion fluxes, enhancement of lysosomal enzyme release, superoxide anion production, chemotaxis, and chemokinesis.
LTB4 is metabolized in leukocytes and hepatocytes to less active 20-hydroxy and 20-carboxy LTB4 and then further oxidized to ω-carboxy dinor LTB4 and ω-carboxy tetranor-LTB₃. It is not excreted in the urine.
Delivery information: Kit contains antibody, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover, Ellman's reagent, tracer and antiserum dye.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty