Would you like to log in again?
Cholesterol Cell-Based Detection Assay Kit, Cayman Chemical Company
Supplier: Cayman Chemical
10009779-192
75816-742EA 372.6 USD InStock
75816-742
Cholesterol Cell-Based Detection Assay Kit, Cayman Chemical Company
Assays Cellular Assays
A 192-well assay for measurement of cholesterol.
  • Detect cholesterol uptake and localization within cultured cells
  • Employs filipin III, a probe that fluoresces in the presence of cholesterol
  • An inhibitor of cholesterol trafficking is included as a positive control
The mechanism for the movement of cholesterol from intracellular sites to their ultimate cellular destination is an unresolved question of fundamental importance to cell biology and medicine. Thus, defining mechanisms of intracellular cholesterol transport and identifying the cellular factors involved are therefore of great interest. Cayman’s Cholesterol Cell-Based Detection Assay includes filipin III, fixative, and wash buffer in a ready to use format. It provides a simple fluorometric method to study mechanisms and biological factors that regulate cholesterol metabolism or movement within cells. A cholesterol trafficking inhibitor, U-18666A, is included as a positive control.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty