Would you like to log in again?
XTT Cell Proliferation Assay Kits, Cayman Chemical
Supplier: Cayman Chemical
10010200-96 10010200-480
75816-752EA 74.82 USD InStock
75816-752 75816-750
XTT Cell Proliferation Assay Kits, Cayman Chemical
Assays Cellular Assays
A colorimetric, non-radioactive assay for assessing cell viability and proliferation. Tetrazolium salt, XTT, is reduced to a soluble purple formazan by trans-plasma membrane electron transport from NADH via an electron mediator.
  • Detect 500 to 100000 cells per well
  • Allows investigators to screen drug candidates involved in cell cycle regulation
Cell proliferation assays are widely used to assess cell cycle regulatory factors such as growth factors, cytokines, mitogens, and drugs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty