Would you like to log in again?
Triglyceride Colorimetric Assay Kit, Cayman Chemical
Supplier: Cayman Chemical
10010303-96
75816-756EA 307.72 USD InStock
75816-756
Triglyceride Colorimetric Assay Kit, Cayman Chemical
Assays Cellular Assays
A simple, reproducible, and sensitive tool for assaying triglycerides in plasma, serum, cell lysates, and tissue homogenate samples.
  • Measure TG levels in plasma, serum, cell lysates, and tissue homogenates
  • Assay 26 samples in triplicate and 40 samples in duplicate
  • Assay Range: 3.125-200 mg/dl
  • Incubation: 15 minutes | Read: Colorimetric at 530-550 nm
The measurement of triglyceride levels, in conjunction with other lipid assays, are useful in the diagnosis of primary and secondary hyperlipoproteinemia, dyslipidemia, and triglyceridemia. Cayman’s Triglyceride Assay Kit provides a simple, reproducible, and sensitive tool for assaying triglycerides in plasma, serum, cell lysates, and tissue homogenate samples. The assay is initiated with the enzymatic hydrolysis of the triglycerides by lipase to produce glycerol and free fatty acids. The glycerol released is subsequently measured by a coupled enzymatic reaction system with a colorimetric readout at 540 nm.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty