Would you like to log in again?
S-Glutathionylated Protein Detection Kit, Cayman Chemical Company
Supplier: Cayman Chemical
10010721-1
75816-760EA 434.82 USD InStock
75816-760
S-Glutathionylated Protein Detection Kit, Cayman Chemical Company
Assays Cellular Assays
A convenient method for the direct visualization of S-glutathionylated proteins in whole (permeabilized) cells.
  • Direct visualization of S-glutathionylated proteins in whole (permeabilized) cells by flow cytometry and microscopy
  • Visualization using colorimetric or fluorescence detection
  • Reagents provided to test up to 30 samples
Glutathione (GSH) is involved in maintenance of protein sulfhydryl reduction status. PSSG may modify the activity of a large number of cell proteins, including metabolic, structural, cytoskeletal, and signaling proteins. Cayman’s PSSG provides a convenient method for the direct visualization of S-glutathionylated proteins in whole (permeabilized) cells by flow cytometry and microscopy as well as avidin overlay analysis. This cell-based assay starts with the modification of protein free-thiols groups followed by enzymatic cleavage of any PSSG adducts present in the sample. Biotinylation of the newly-formed protein free-thiols provides the basis for visualization using streptavidin-based colorimetric or fluorescence detection. Reagents are provided to test three sets of 10 samples (most convenient) or up to thirty samples total at once if desired.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty