Would you like to log in again?
Cortisol Express ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical
500370-96 500370-480
75816-782EA 1283.2 USD InStock
75816-782 75816-784
Cortisol Express ELISA Kit, Cayman Chemical Company
Assays ELISAs
A fast, convenient assay format for measurement of cortisol.
  • Measure cortisol in urine, plasma, and other sample matrices
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure cortisol levels down to 110 pg/ml
  • Rapid assay; get results in under four hours
Cortisol Express ELISA is a competitive assay that permits the rapid measurement of cortisol from biological samples, requiring only a two-hour incubation and one-hour development times. This ELISA offers the convenience of a fast assay while maintaining sensitivity.
Delivery information: Kit contains monoclonal antibody, AChE tracer, standard, ELISA and wash buffer, polysorbate 20, coated plate, cover sheet, Ellman's reagent, tracer and antiserum dye.
Caution: Due to an approximate 15% cross-reactivity with dexamethasone, this kit may not be suitable for use with samples that contain high concentrations of dexamethasone, such as those collected during a dexamethasone suppression test.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty