Would you like to log in again?
Resolvin D1 ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical
500380-480 500380-96
75816-788EA 608.88 USD InStock
75816-788 75816-786
Resolvin D1 ELISA Kit, Cayman Chemical Company
Assays ELISAs
For the quantification of RvD1.
  • Detect RvD1 in multiple sample types
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure RvD1 levels down to 15 pg/mL
  • Incubation: 18 hours | Development: 90 – 120 minutes | Read: Colorimetric at 405 – 420 nm
  • Midpoint: 50% B/B0: 50 – 100 pg/mL
Cayman’s Resolvin D1 ELISA Kit is a competitive assay that can be used for quantification of RvD1. Due to the number and variation of potential sample types, this assay has been validated in Cayman’s ELISA Buffer (Item No. 400060) diluted to 1X (0.1 M potassium phosphate containing 0.1% BSA, 0.1% sodium chloride, 1 mM EDTA, and 0.01% sodium azide) and DMEM containing phenol red and 10% FBS.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty