Would you like to log in again?
Oxytocin ELISA Kit, Cayman Chemical
Supplier: Cayman Chemical
500440-96 500440-480
75816-800EA 2140.08 USD InStock
75816-800 75816-802
Oxytocin ELISA Kit, Cayman Chemical
Assays ELISAs
For the quantification of oxytocin in plasma.
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure oxytocin levels down to 20 pg/mL
  • Incubation: 18 hours | Development: 90 – 120 minutes | Read: Colorimetric at 405 – 420 nm
  • Midpoint: 50% B/B0: 50 – 100 pg/mL
Oxytocin is a hypothalamic peptide hormone that is stored in the posterior pituitary gland. It is released into the bloodstream during parturition and lactation and is also involved in other social and sexual behaviors, neuropsyciatric disorders, and the maintenance of water and sodium homeostasis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty