Would you like to log in again?
Creatinine (urinary) Colorimetric Assay Kit, Cayman Chemical Company
Supplier: Cayman Chemical
500701-96 500701-480
75816-810EA 132 USD InStock
75816-810 75816-808
Creatinine (urinary) Colorimetric Assay Kit, Cayman Chemical Company
Assays Protein Assays
For the measurement of creatinine levels in urine
  • Measure creatinine levels in urine
  • Assay 40 samples in duplicate
  • Measure creatinine levels down to 0.1 mg/dL
  • Plate-based colorimetric measurement (490 – 500 nm)
Creatine is synthesized in kidney, liver, and pancreas and transported to muscle and brain where it is phosphorylated to phosphocreatine. Some free creatine in muscle is converted to creatinine. The amount of creatinine produced is proportional to an individual’s muscle mass. In the absence of renal disease, the excretion rate of creatinine in an individual is relatively constant. Thus, urinary creatinine levels are commonly used as an index of standardization for a variety of other tests. Measurement of creatinine clearance is also useful in detecting renal disease and estimating the extent of impairment of renal function.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty