Would you like to log in again?
Anti-Ovalbumin IgG1 (Mouse) ELISA Kit, Cayman Chemical
Supplier: Cayman Chemical
500830-96
75816-812EA 628.67 USD InStock
75816-812
Anti-Ovalbumin IgG1 (Mouse) ELISA Kit, Cayman Chemical
Assays ELISAs
For the measurement of anti-ovalbumin of the IgG1 isotype in mouse plasma and serum.
  • Measure mouse anti-OVA IgG1 in mouse plasma, serum, or BAL fluids
  • Employs an affinity-purified mouse anti-OVA IgG1 polyclonal antibody standard for accurate results
  • Assay Range: 1.56 – 200 ng/mL with an LLOQ of 1.56 ng/mL
  • Rapid assay; get results in under 4 hours
Cayman's Ovalbumin IgG1 (mouse) ELISA Kit is an immunometric (i.e., “sandwich”) assay which can be used to measure anti-ovalbumin of the IgG1 isotype in mouse plasma and serum without prior sample purification.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty