Would you like to log in again?
Human Therapeutic IgG4 ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical
500920-96
75816-820EA 963.44 USD InStock
75816-820
Human Therapeutic IgG4 ELISA Kit, Cayman Chemical Company
Assays ELISAs
A generic assay for the quantitation of therapeutic human IgG4 antibodies in monkey serum or plasma.
  • Specifically detect human IgG4 antibodies in monkey plasma or serum
  • Not cross reactive to rhesus or cynomolgus immunoglobulins
  • Useful for pharmacokinetic analysis of therapeutic human IgG4 antibodies in monkeys
  • Useful for detecting human IgG4 in mouse or rat plasma or serum
Human therapeutic antibodies have become increasingly common components of early drug discovery and development portfolios in the pharmaceutical and biotech industries. As part of preclinical toxicology assessment of these agents, they are routinely tested in non-human primates, primarily in rhesus or cynomolgus monkeys. In order to assess the pharmacokinetics of the human antibodies in monkey serum, it is necessary to have an assay that is capable of distinguishing the experimentally-introduced human IgG from the endogenous monkey IgG. Cayman’s IgG4 (human) Pharmacokinetic ELISA Kit is an immunometric assay which can be used to measure human IgG4 in monkey or rodent plasma and serum without prior sample purification. The standard curve spans the range of 7.8-500 ng/ml, with an LLOQ of 7.8 ng/ml. The assay has been validated in serum from cynomolgus monkeys.
Delivery information: Kit contains: Standard, HRP-Conjugate, Buffers, Reagents, and precoated 96-well Plate
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty