Would you like to log in again?
PAD4 Autoantibody ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical
500930-96
75816-822EA 712.03 USD InStock
75816-822
PAD4 Autoantibody ELISA Kit, Cayman Chemical Company
Assays ELISAs
An immunometric for the measurement of anti-PAD4 autoantibodies.
  • Measure anti-PAD4 autoantibodies of any isotype (IgM, IgG, IgA)
  • For use in human plasma and serum (no need to purify samples)
  • Lower Limit of quantification is 31.3 U/ml
PAD4 is a guanidino-modifying enzyme which catalyzes the conversion of specific arginine residues to citrulline in a calcium-dependent manner. PAD4 activity is increased in rheumatoid arthritis (RA), producing citrulline-containing proteins (CCP) that induce the formation of arthritogenic autoantibodies.
In addition to anti-CCP autoantibodies, 23 to 45% of RA patients also produce autoantibodies specific for the PAD4 enzyme itself. PAD4 Autoantibody ELISA Kit is an immunometric assay which can be used to measure anti-PAD4 autoantibodies of any isotype (IgM, IgG, and IgA) in human plasma and serum without prior sample purification.
Affinity-purified anti-PAD4 antibody isolated from the plasma of a patient with RA is used as the standard. One unit is approximately equal to 1 ng of anti-PAD4 Ig protein.
Delivery information: Kit contains HRP conjugate, plate, ELISA standard, wash buffer concentrate, non-fat dry milk assay reagent, ELISA buffer concentrate, polysorbate 20, substrate and stop solution, cover sheet.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty