Would you like to log in again?
Thromboxane B₂ ELISA Kit, Cayman Chemical
Supplier: Cayman Chemical
501020-480 501020-96
75816-832EA 478.66 USD InStock
75816-832 75816-830
Thromboxane B₂ ELISA Kit, Cayman Chemical
Assays ELISAs
For the quantification of TXB2 in urine, serum, tissue culture supernatants, and other sample matrices.
  • Measure TXB2 in multiple sample types
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure TXB2 levels down to 5 pg/ml
  • Incubation: 18 hours | Development: 90-120 minutes | Read: Colorimetric at 405-420 nm
Thromboxane A2 (TXA2) is produced from arachidonic acid by many cells and causes irreversible platelet aggregation and contraction of vascular and bronchial smooth muscle. Like most lipid mediators, TXA2 is not a circulating hormone. It is formed in response to local stimuli and exerts its effects within a short distance of its biosynthesis. TXA2 is rapidly hydrolyzed non-enzymatically to form TXB2, which is then quickly metabolized (t½ = 5-7 minutes) to urinary metabolites for clearance by the kidneys. Because of the transient nature of this compound it is difficult to accurately measure circulating levels in whole animal experimental models. In fact, it has been shown that plasma and urine levels of TXB2 are primarily due to ex vivo platelet activation and intra-renal production, respectively. Therefore, measurement of TXB2 metabolites, such as 11-dehydro TXB2, in urine and plasma may give more accurate estimates of in vivo TXA2 production. TXB2 measurement is better suited for samples that are unlikely to undergo extensive metabolism such as perfusates, lavage samples, tissue/cell culture, etc.
Delivery information: Kit contains: Antiserum, Tracer, Standard, Buffers, Reagents, and Plate(s).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty