Would you like to log in again?
Vitamin D ELISA Kit, Cayman Chemical
Supplier: Cayman Chemical
501050-480 501050-96
75816-842EA 779.28 USD InStock
75816-842 75816-840
Vitamin D ELISA Kit, Cayman Chemical
Assays ELISAs
Sensitive ELISA for Vitamin D quantification.
  • Measure vitamin D in plasma or serum
  • Detect the metabolically stable forms 25(OH)D3 and 25(OH)D2
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure vitamin D levels down to 0.5 ng/ml
  • Read: Colorimetric at 405 to 420 nm
The primary synthesis of functional vitamin D begins in the skin, where a cholesterol by-product acted upon by UV light forms vitamin D3. Vitamin D3 is carried through the bloodstream, attached to the vitamin D binding protein, to the liver where it is converted to 25-hydroxy vitamin D3 (25(OH)D3). This product is then transported to the kidney where it is further converted to 1,25-dihydroxycholecalciferol (1,25(OH)2D3), the biologically active form. Active 1,25(OH)2D3 is very short-lived and is rapidly metabolized to its deactivated forms.
Vitamin D Immunoassay, accurate Vitamin D measurement in plasma or serum. Vitamin D ELISA Kit is a competitive assay that can be used for Vitamin D quantification in plasma or serum. Because of the short half-life of the biologically active 1,25(OH)D form, this Vitamin D assay primarily detects the more metabolically stable forms, 25(OH)D3 and 25(OH)D2.
Accurate detection of these forms requires that they be displaced from the vitamin D binding protein prior to measurement. A simple, acetone-based purification protocol is included as the initial procedure of the Vitamin D assay kit for this purpose.
Delivery information: Kit contains antibody, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover, Ellman's reagent, tracer and antiserum dye.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty