Would you like to log in again?
Leukotriene E4 ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical
501060-96 501060-480
75816-844EA 1739.36 USD InStock
75816-844 75816-846
Leukotriene E4 ELISA Kit, Cayman Chemical Company
Assays ELISAs
For the quantification of LTE4 in urine.
  • Measure LTE4 in urine, with no purification required
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure LTE4 levels down to 25 pg/mL
  • Incubation: 18 hours | Development: 90 – 120 minutes | Read: Colorimetric at 405 – 420 nm
Leukotriene E4 (LTE4) is a product of the 5-lipoxygenase (5-LO) pathway in activated mast cells, eosinophils, and monocytes. LTA4, the primary 5-LO metabolite, is rapidly converted to LTC4 and sequentially to LTD4 and LTE4. While plasma levels of LTC4 are virtually undetectable, exogenously administered LTC4 is recovered in the urine as LTE4 (5 – 13%) and two prominent oxidized metabolites resulting from several cycles of β-oxidation. Cayman’s LTE4 ELISA Kit is a competitive assay developed for the measurement of LTE4 from urine without a purification step. The assay has a range of 7.8 – 1000 pg/mL and a sensitivity (80% B/B0) of approximately 25 pg/mL. This “dilute and go” format has been used successfully to demonstrate significant and reproducible changes in LTE4 levels in the urine of asthmatic patients after antigen challenge. If your samples require additional purification prior to analysis, Cayman’s Cysteinyl Leukotriene Affinity Sorbent can be used for this purpose.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty