Would you like to log in again?
Leukotriene C₄ ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical
501070-96 501070-480
75816-848EA 2038.95 USD InStock
75816-848 75816-850
Leukotriene C₄ ELISA Kit, Cayman Chemical Company
Assays ELISAs
For the quantification of LTC₄ in culture media and other sample matrices.
  • Measure LTC₄ in tissue culture medium
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure LTC₄ levels down to 40 pg/mL
  • Incubation: 18 hours | Development: 90 – 120 minutes | Read: Colorimetric at 405 – 420 nm
Leukotrienes (LTs) are a group of acute inflammatory mediators derived from arachidonic acid in eosinophils, mast cells, and macrophages. Their biosynthesis proceeds via the 5-lipoxygenase (5-LO) pathway in which 5-LO converts arachidonic acid into LTA₄ with 5(S)-HpETE as an intermediate. The conjugation of glutathione to LTA₄ results in the formation of LTC₄, which is then rapidly metabolized to LTD₄ and LTE₄. This metabolism is essentially complete within 10 minutes in the human lung. Cayman’s LTC₄ ELISA Kit is a competitive assay that can be used for quantification of LTC₄ in culture media and other sample matrices. The assay has a range from 17.1 – 5000 pg/mL and a sensitivity (80% B/B₀) value of approximately 40 pg/mL.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty