Would you like to log in again?
Prostaglandin I Metabolite ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical
501100-480 501100-96
75816-856EA 424.37 USD InStock
75816-856 75816-854
Prostaglandin I Metabolite ELISA Kit, Cayman Chemical Company
Assays ELISAs
For the quantification of PGIM in urine and other sample matrices.
  • Measure PGIMs in urine
  • Measure PGIM down to 120 pg/mL
  • Convenient overnight format
Prostaglandin I (Prostaglandin I2; PGI2) is a potent vasodilator and inhibitor of platelet aggregation that is formed from arachidonic acid primarily in the vascular endothelium and renal cortex by sequential activities of cyclooxygenase (COX) and PGI2 synthase. It is non-enzymatically hydrated to 6-keto PGF (t½ = 2-3 minutes), and then quickly converted to numerous downstream metabolites, collectively known as prostaglandin I metabolites or PGIM, (t½= 30 minutes). Although 6-keto PGF is commonly measured in plasma and urine as an estimate of PGI2 synthesis, it should be noted that there may be more than one source of PGI2 in these samples. For instance, urinary concentrations of 6-keto PGF are confounded by the fact that only a portion originates from plasma and the remainder is produced by the kidney. These problems are circumvented by measuring downstream PGIM rather than 6-keto PGF as an indicator of systemic PGI2 production. Cayman's PGIM ELISA Kit is a competitive assay that can be used for quantification of PGIM in urine and other sample matrices. The assay has a range from 39 – 5000 pg/mL and a sensitivity (80% B/B0) of approximately 120 pg/mL.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty