Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Corticosterone ELISA Kit, Cayman Chemical Company

Supplier: Cayman Chemical
501320-480 501320-96
75816-864EA 413.06 USD InStock
75816-864 75816-862
Corticosterone ELISA Kit, Cayman Chemical Company
Assays ELISAs
A competitive assay for the quantification of corticosterone in serum and rodent feces.

  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure corticosterone levels down to 30 pg/mL
  • Incubation: Overnight | Development: 90 – 120 minutes | Read: Colorimetric at 405 – 420 nm
  • Mid-point: 50% B/B0: 130 – 180 pg/mL

Corticosterone is a glucocorticoid produced by the adrenal cortex in response to ACTH (adrenocorticotropic hormone), and is the precursor to aldosterone. The production of glucorcorticoids is increased by stress; therefore, corticosterone can be used as a biomarker of stress. Plasma corticosterone levels have a circadian variation, and may be important in the regulation of the sleep-wake cycle. Cayman’s Corticosterone ELISA Kit is a competitive assay that can be used for quantification of corticosterone in serum and rodent feces.

Delivery information: Kit contains: Antiserum, Tracer, Standard, Buffers, Reagents, and Plate.

Packaging: Kit contains: Antiserum, Tracer, Standard, Buffers, Reagents, and Plate

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty