Would you like to log in again?
Prostaglandin D₂ Express ELISA Kit, Cayman Chemical
Supplier: Cayman Chemical
512041-480 512041-96
75816-866EA 631.52 USD InStock
75816-866 75817-276
Prostaglandin D₂ Express ELISA Kit, Cayman Chemical
Assays ELISAs
A competitive assay that can be used for quantification of PGD2.
  • Measure PGD2 in multiple sample types
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure PGD2 levels down to 350 pg/mL
  • Rapid assay; get results in under 4 hours
  • Midpoint: 50% B/B0: 800 – 1,500 pg/mL
Prostaglandin D2 (PGD2) is biosynthesized in the brain by soluble, glutathione-independent lipocalin-type PGD2 synthase. PGD2 accumulates in the cerebrospinal fluid (CSF), where it induces physiologic sleep in rats and humans. PGD2 is also synthesized in mast cells and leukocytes by a cellular, myeloid-type, glutathione-dependent PGD synthase. This PGD2 which is formed in the intracellular and vascular compartments is rapidly metabolized to 11β-PGF and other metabolites. Measurement of the parent eicosanoid PGD2 is appropriate in cell culture lysates and in CSF, where concentrations of several hundred pg/ml have been measured. Cayman’s PGD2 Express ELISA Kit is a competitive assay that can be used for quantification of PGD2 in cell culture lysates and enzymatic reactions. This ELISA offers the convenience of a fast assay with a two hour incubation time and one hour development time, while still achieving an assay range of approximately 117.2 – 15,000 pg/mL and a sensitvity (80% B/B0) of approximately 350 pg/mL.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty