Would you like to log in again?
Prostaglandin E₂ ELISA Kit - Monoclonal, Cayman Chemical
Supplier: Cayman Chemical
514010-480 514010-96
75816-868EA 1720.05 USD InStock
75816-868 75816-870
Prostaglandin E₂ ELISA Kit - Monoclonal, Cayman Chemical
Assays ELISAs
For quantification of PGE2 in plasma, urine, and culture media samples.
  • Measure PGE2 in multiple sample types
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure PGE2 levels down to 15 pg/ml
  • Incubation: 18 hours | Development: 60-90 minutes | Read: Colorimetric at 405-420 nm
Prostaglandin E2 (PGE2) is a primary product of arachidonic acid metabolism in many cells. Like most eicosanoids, it does not exist preformed in any cellular reservoir. When cells are activated or exogenous free arachidonate is supplied, PGE2 is synthesized de novo and released into the extracellular space. In vivo, PGE2 is rapidly converted to an inactive metabolite (13,14-dihydro-15-keto PGE2) by the prostaglandin 15-dehydrogenase pathway. The half-life of PGE2 in the circulatory system is approximately 30 seconds and normal plasma levels are 3-12 pg/ml. Our PGE2 ELISA has been validated for use with urine, plasma, and culture media samples. In general, urine and culture media samples can be diluted, if necessary, and added directly to the assay well. Plasma samples should be purified prior to use. Because of the rapid metabolism of PGE2, the determination of in vivo PGE2 biosynthesis is often best accomplished by the measurement of PGE2 metabolites.
Delivery information: Kit contains: Antibody, tracer, standard, buffers, reagents, and plate(s).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty