Would you like to log in again?
Prostaglandin E Metabolite ELISA Kit, Cayman Chemical
Supplier: Cayman Chemical
514531-96 514531-480
75816-878EA 431.46 USD InStock
75816-878 75816-876
Prostaglandin E Metabolite ELISA Kit, Cayman Chemical
Assays ELISAs
Competitive ELISA for quantitation of a primary metabolite of PGE2 in biological samples.
  • Converts major urinary and plasma metabolites of PGE2 to a single, stable derivative that is easily quantified
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure PGEM levels down to 2 pg/mL
  • Incubation: 18 hours | Development: 60 – 90 minutes | Read: Colorimetric at 405 – 420 nm
  • Midpoint: 50% B/B0: 90 – 140 pg/mL
Prostaglandin E2 (PGE2) is produced by a variety of cell types which, in general, do not contain the enzymes required for metabolism of PGE2. Thus, cultured endothelial cells or osteoblasts will release PGE2 into the culture medium, where it will accumulate without appreciable metabolism. The direct assay of PGE2 from the medium is a good way to measure PGE2 production from these cells. PGE2 is rapidly converted in vivo to its 13,14-dihydro-15-keto metabolite, with more than 90% of circulating PGE2 cleared by a single passage through the lungs. Unfortunately, this metabolite is not chemically stable and undergoes a variable amount of degradation to PGA products. For this reason, blood, urine, or other samples from whole animals or humans often contain very little intact PGE2, and measurement of the metabolites is necessary to provide a reliable estimate of actual PGE2 production. Cayman’s PGE Metabolite assay was developed as a method of converting 13,14-dihydro-15-keto PGA2 and 13,14-dihydro-15-keto PGE2 to a single, stable derivative that could be easily quantified by ELISA. This assay is therefore the method of choice if the samples in question have undergone extensive metabolism prior to collection.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty