Would you like to log in again?
Prostaglandin F ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical
516011-480 516011-96
75816-886EA 447.02 USD InStock
75816-886 75816-884
Prostaglandin F ELISA Kit, Cayman Chemical Company
Assays ELISAs
For the quantification of PGF in tissue culture supernatants and other sample matrices.
  • Measure PGF in multiple sample types
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure PGF levels down to 10 pg/mL
  • Incubation: 18 hours | Development: 90 – 120 minutes | Read: Colorimetric at 405 – 420 nm
PGF is one of the five primary PGs derived enzymatically directly from the endoperoxide PGH2. The majority of the functional roles ascribed to it relate to fertility, pregnancy, and parturition. Like all of the primary PGs, PGF has a very short half-life in the general circulation. The plasma concentration of PGF in humans is <10 pg/mL, and probably no more than 1 – 2 pg/mL. Cayman’s PGF ELISA Kit is a competitive assay that can be used for the quantification of PGF in tissue culture supernatants and other sample matrices.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty