Would you like to log in again?
11β-Prostaglandin F ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical
516521-96 516521-480
75816-898EA 358.62 USD InStock
75816-898 75816-896
11β-Prostaglandin F ELISA Kit, Cayman Chemical Company
Assays ELISAs
A sensitive assay for the detection of 11β-PGF.
  • Detect 11β-PGF, the primary metabolite of PGD2, in plasma or urine
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure 11β-PGF levels down to 5 pg/mL
  • Incubation: 18 hours | Development: 60 – 90 minutes | Read: Colorimetric at 405 – 420 nm
  • Midpoint: 50% B/B0: 25 – 45 pg/mL
11β-PGF is the primary plasma metabolite of PGD2 in vivo, the levels of which can increase from 6 pg/mL in a normal healthy volunteer to 490 ng/mL in patients with systemic mastocytosis. The normal human urinary excretion of 11β-PGFis about 11 ng/mmol creatinine (~400 ng/24 hr), which is increased nearly 3-fold upon allergen-induced bronchoconstriction in asthmatics. Cayman’s 11β-PGF Assay is a competitive assay that provides accurate measurements of 11β-PGF within the range of 1.6 – 1,000 pg/mL.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty