Would you like to log in again?
13,14-dihydro-15-keto Prostaglandin F ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical
516671-96 516671-480
75816-900EA 1468.78 USD InStock
75816-900 75816-902
13,14-dihydro-15-keto Prostaglandin F ELISA Kit, Cayman Chemical Company
Assays ELISAs
This kit is used for the measurement of 13,14-dihydro-15-keto prostaglandin F2α in plasma.
  • Detect 13,14-dihydro-15-keto PGF, a stable metabolite of PGF
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure 13,14-dihydro-15-keto PGF levels down to 16.3 pg/ml
  • Reads colorimetric at 405 to 420 nm
PGF is rapidly metabolized to 13,14-dihydro-15-keto PGF in vivo, by the enzymes 15-hydroxy PG dehydrogenase and D13-reductase. Measurement of 13,14-dihydro-15-keto PGF in plasma can be used as a marker of the in vivo production of PGF.
13,14-dihydro-15-keto PGF ELISA is a competitive assay that provides measurements of 13,14-dihydro-15-keto PGF within the range of 2.3 to 5000 pg/ml. The assay has been validated for the measurement of 13,14-dihydro-15-keto PGF in plasma, a common matrix for measurement of this PGF metabolite.
Delivery information: Kit contains antiserum, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover, Ellman's reagent, tracer and antiserum dye.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty