Would you like to log in again?
Latanoprost ELISA Kit, Cayman Chemical
Supplier: Cayman Chemical
516811-480 516811-96
75816-910EA 358.62 USD InStock
75816-910 75816-908
Latanoprost ELISA Kit, Cayman Chemical
Assays ELISAs
A sensitive detection method for measuring the free acid form of latanoprost. It also detects the isopropyl ester form of latanoprost, but with less sensitivity.
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure latanoprost levels down to 15.5 pg/ml
  • Read: Colorimetric at 405 to 420 nm
Latanoprost is 13,14-dihydro-17-phenyl-18,19,20-trinor prostaglandin F2α isopropyl ester (13,14-dihydro-17-phenyl-18,19,20-trinor PGF2α isopropyl ester), an F-series prostaglandin analog which has been approved for use as an ocular hypotensive drug.
Prostaglandin esters and other similar derivatives act as prodrugs which are converted to the active free acid form by an esterase/amidase activity in ocular tissues. The free acid form of latanoprost (latanoprost free acid (Lat-FA), or PhXA 85) is 200 times more potent than the isopropyl ester as an FP receptor ligand tested using the recombinant human FP receptor.
Although the exact mechanism of the intraocular hypotensive effect of prostaglandins is unknown, it correlates very closely with the FP receptor ligand binding affinity of FP receptor agonists.
Latanoprost ELISA is a sensitive detection method for measuring the free acid form of Latanoprost. It also detects the isopropyl ester form of Latanoprost, but with less sensitivity. The assay is most appropriate for use when only one of the two forms is present. Samples containing mixtures of both the isopropyl ester and free acid should be purified and the two compounds separated prior to ELISA analysis.
Delivery information: Kit contains antiserum, tracer, two ELISA standards, ELISA and wash buffer concentrate, polysorbate 20, plate and cover sheet, Ellman's reagent, tracer and antiserum dye.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty