Would you like to log in again?
Estriol ELISA Kit, Cayman Chemical
Supplier: Cayman Chemical
582281-480 582281-96
75816-932EA 1225.67 USD InStock
75816-932 75816-934
Estriol ELISA Kit, Cayman Chemical
Assays ELISAs
Estriol ELISA kit is used for the quantification of estradiol in plasma and other sample matrices.
  • Measure estradiol (17β-estradiol) in cell culture medium, plasma, and serum
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure estradiol levels down to four pg/ml
  • Read: Colorimetric at 405 to 420 nm
Estradiol is the major estrogen secreted by the premenopausal ovary. Estrogens direct the development of the female genotype in embryogenesis and at puberty. In addition, estradiol is an important luteolytic agent in humans.
The majority of the estriol synthesized in the later stages of pregnancy originates from fetal dehydroepiandrosterone sulfate (DHEAS) and serves as a direct marker of fetal adrenal gland activity. However, saliva contains primarily unbound and unconjugated estriol, the biologically active form of the hormone, and therefore is currently a commonly used biological fluid for monitoring estriol levels. Plasma levels of estriol in males and non-pregnant females is less than 2 ng/ml.
Delivery information: Kit contains antiserum, tracer, standard, ELISA and wash buffer, polysorbate 20, coated plate and cover sheet, Ellman's reagent, tracer and antiserum dye.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty