Would you like to log in again?
Progesterone ELISA Kit, Cayman Chemical
Supplier: Cayman Chemical
582601-96 582601-480
75816-936EA 1060.17 USD InStock
75816-936 75816-938
Progesterone ELISA Kit, Cayman Chemical
Assays ELISAs
For the quantification of progesterone in plasma and other sample matrices.
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure progesterone levels down to 10 pg/mL
  • Rapid assay; get results in under 4 hours
  • Midpoint: 50% B/B0: 40 – 90 pg/mL
Progesterone, along with pregnenolone, is the biosynthetic precursor of all other steroid hormones. The main function of progesterone is to prepare the uterine lining for implantation of a fertilized ovum and to maintain pregnancy. Measurement of serum or plasma progesterone levels are used as an index to monitor ovulation and investigate luteal function.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty