Would you like to log in again?
11-keto Testosterone ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical
582751-96 582751-480
75816-944EA 1728.84 USD InStock
75816-944 75816-946
11-keto Testosterone ELISA Kit, Cayman Chemical Company
Assays ELISAs
For the quantification of 11-keto testosterone in plasma and other sample matrices.
  • Detect the primary androgenic steroid in fish
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure 11-keto testosterone levels down to 1.3 pg/mL
  • Incubation: 18 hours | Development: 90 – 120 minutes | Read: Colorimetric at 405 – 420 nm
  • Mid-point: 50% B/B0: 2 – 10 pg/mL
11-KT is a second key androgenic steroid found in fish. It occurs in males together with testosterone in amounts which vary from less than 1 ng/mL to as much as 50 – 100 ng/mL, depending on the species and the stage of the reproductive cycle. Cayman's 11-keto Testosterone ELISA Kit is a competitive assay tha can be used for quantification of 11-keto testosterone in plasma and other sample matrices.
Delivery information: Kit contains: Antiserum, Tracer, Standard, Buffers, Reagents, Plate, and Dye.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty