Would you like to log in again?
Endothelin ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical
583151-96 583151-480
75816-948EA 2147.96 USD InStock
75816-948 75816-950
Endothelin ELISA Kit, Cayman Chemical Company
Assays ELISAs
A sensitive assay for the specific analysis of endothelin in serum, plasma, urine, or cell culture medium.
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Read: Colorimetric at 405 to 420 nm
The endothelin peptide family consists of three isoforms, ET-1 (corresponding to the initially isolated and most predominant isoform), ET-2, and ET-3. ET-1 is a 21 amino acid peptide and is one of the most potent vasoconstritors currently known. ET-2 displays similar pharmacology to ET-1, whereas ET-3 is a weak vasoconstrictor but more potent inhibitor of platelet aggregation.
Inter- and intra-assay CV’s of less than 10% may be achieved at most concentrations. This assay offers sensitive and specific analysis of endothelin in serum, plasma, urine, or cell culture media.
Delivery information: Kit contains plate, AChE-Fab' conjugate, standard, wash and ELISA buffer concentrate, polysorbate 20, cover sheet, Ellman's reagent, human plasma, and serum.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty