Would you like to log in again?
Interleukin-1α (human) ELISA Kit, Cayman Chemical
Supplier: Cayman Chemical
583301-480 583301-96
75816-954EA 528.07 USD InStock
75816-954 75816-952
Interleukin-1α (human) ELISA Kit, Cayman Chemical
Assays ELISAs
An immunometric assay for the measurement of IL-1α.
  • Measure IL-1α from human plasma, serum, synovial fluid, and other sample matrices
  • Incubation: 18 hours
  • Development: 60 to 90 minutes
  • Read: Colorimetric at 405 to 420 nm
Cayman’s IL-1α (human) ELISA is an immunometric ( sandwich) ELISA that permits IL-1α measurements within the range of 3.9 to 250 pg/ml, typically with a limit of detection of 3.9 pg/ml.
Delivery information: Kit contains: ELISA Strip Plate, AChE Fab' Conjugate, Standard, ELISA Buffer Concentrate, Wash Buffer Concentrate, Polysorbate 20, 96-Well Cover Sheet, Ellman's Reagent, Human Plasma, and Non-specific Mouse Serum
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty