Would you like to log in again?
Interleukin-6 (Mouse) ELISA Kit, Cayman Chemical
Supplier: Cayman Chemical
583371-96
75816-960EA 478.66 USD InStock
75816-960
Interleukin-6 (Mouse) ELISA Kit, Cayman Chemical
Assays ELISAs
Enzyme immunometric assay for the measurement of mouse IL-6.
  • Measure IL-6 from mouse plasma, serum, and other sample matrices
  • Rapid assay
  • Get results in under 3 hours
Interleukin-6 (IL-6) is produced by monocytes, macrophages, and T cells and is invovled in many defense mechanisms including the immune response, hematopoiesis, bone metabolism, and acute phase reactions.
Cayman’s IL-6 (mouse) ELISA Kit is an immunometric (sandwich) ELISA that permits IL-6 measurements within the range of 23.4 to 1500 pg/ml, with an LLOQ of approximately 23.4 pg/ml.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty