Would you like to log in again?
Substance P ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical
583751-96 583751-480
75816-964EA 536.23 USD InStock
75816-964 75816-962
Substance P ELISA Kit, Cayman Chemical Company
Assays ELISAs
For the measurement of Substance P.
  • Measure substance P in multiple sample matrices from multiple species
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure substance P levels down to 8 pg/mL
  • Incubation: 18 hours | Development: 90 – 120 minutes | Read: Colorimetric at 405 – 420 nm
  • Midpoint: 50% B/B0: 20 – 50 pg/mL
Substance P is a bioactive 11-amino acid peptide first isolated in 1931 from brain and intestine. The peptide is involved in many physiological processes including pain modulation, smooth muscle contraction, blood pressure control, kidney function, and water homeostasis. Substance P is widely distributed in numerous tissues and body fluids including the central and peripheral nervous system, gastrointestinal tract, respiratory tract, visual system, and circulatory system.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty