Would you like to log in again?
Methionine Sulfoxide Immunoblotting Kit, Cayman Chemical
Supplier: Cayman Chemical
600160-1
75816-974EA 596.83 USD InStock
75816-974
Methionine Sulfoxide Immunoblotting Kit, Cayman Chemical
Assays Immunological Assays
For the immunochemical detection of proteins containing MetO residues by western blotting.
  • Detect proteins containing MetO residues by Western blot
  • Positive control included
Protein MetO is a reversible oxidative modification that occurs by exposure of protein(s) methionine residues to reactive oxygen species (ROS). Methionine oxidation can alter the function(s) of the modified proteins and if not reversed by MetO reductases can be further oxidized to methionine sulfone, an irreversible modification. Cayman’s MetO Immunoblotting Kit contains reagents needed for the immunochemical detection of proteins containing MetO residues by western blotting. MetO-containing samples of interest include those from cell or tissue lysates as well as semi-pure or purified proteins.
Delivery information: Positive control included.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty