Would you like to log in again?
Sulfenylated Protein Cell-Based Detection Kit, Cayman Chemical
Supplier: Cayman Chemical
600320-96
75816-980EA 472.91 USD InStock
75816-980
Sulfenylated Protein Cell-Based Detection Kit, Cayman Chemical
Assays Cellular Assays
For the detection of sulfenic acid-modified proteins in living cells.
  • Contains the cell-permeable probe DAz-2 to detect sulfenic acid-modified proteins in cells
  • Includes EGCG as a positive control
  • Detect by flow cytometry, fluorescence microscopy, or plate reader
Reactive oxygen species (ROS) react with proteins, resulting in protein modification, such as protein sulfenylation through the reversible oxidation of cysteine residues. Redox-sensitive cysteine residues in proteins can function as sensors of ROS and serve as molecular switches, activating or deactivating proteins, following a change in oxidative state. However, the accumulation of proteins with irreversible cysteine oxidation is a hallmark of stress-induced cellular damage associated with diseases like cancer. Cayman’s Sulfenylated Protein Cell-Based Detection Kit employs the cell-permeable and chemoselective DAz-2 probe to detect sulfenic acid-modified proteins in living cells. It can be used to monitor intracellular sulfenylated protein levels in living cells to discriminate between normal and pathological conditions or to help identify new pathways regulated by sulfenic acid formation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty