Would you like to log in again?
Glycolysis Cell-Based Assay Kit, Cayman Chemical Company
Supplier: Cayman Chemical
600450-1
75816-990EA 419.5 USD InStock
75816-990
Glycolysis Cell-Based Assay Kit, Cayman Chemical Company
Assays Cellular Assays
A colorimetric-based method for detecting L-lactate in culture medium
  • Detect L-lactate, the end product of glycolysis
  • Can be adapted to high-throughput screening
  • Measure L-lactate in cell culture supernatant down to 156 µM
Cayman’s Glycolysis Cell-Based Assay Kit provides a colorimetric method for detecting L-lactate, the end product of glycolysis, produced and secreted by cultured cells. In this assay, lactate dehydrogenase catalyzes the reaction between NAD+ and lactate, yielding pyruvate and NADH. The NADH directly reduces a tetrazolium salt (INT) to a colored formazan which absorbs between 490 and 520 nm. The quantity of formazan produced is proportional to the quantity of lactate in the culture medium, and is thus an indirect measurement of glycolysis. This assay can be adapted to high-throughput screening to examine glycolytic regulators in a cell-based system. In addition, the non-invasive sampling leaves the cells intact and thus allows multiplex testing for additional markers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty