Would you like to log in again?
Glucose Uptake Cell-Based Assay Kit, Cayman Chemical Company
Supplier: Cayman Chemical
600470-1
75816-992EA 422.69 USD InStock
75816-992
Glucose Uptake Cell-Based Assay Kit, Cayman Chemical Company
Assays Cellular Assays
A cell-based assay for studying modulators of cellular glucose uptake.
  • Measure glucose uptake by cultured cells
  • Employs 2-NBDG, a fluorescent deoxyglucose analog
  • An inhibitor of glucose transport is included as a control
  • Convenient tool for studying modulators of glucose uptake
Glucose metabolism is a primary source of energy and biomaterials for the maintenance of cell homeostasis. Extra glucose is stored in the muscles and liver as glycogen which is hydrolyzed to glucose and released into the blood when needed. The rate of glucose uptake in cells is dynamic and tightly regulated by hormones and/or growth factors including insulin. Cancer cells exhibit increased glucose uptake and metabolism by aerobic glycolysis in order to support a high rate of proliferation. Chemicals that block glucose uptake by cancer cells have been shown to have anti-cancer effects. Cayman’s Glucose Uptake Cell-based Assay Kit provides a convenient tool for studying modulators of cellular glucose uptake. The kit employs 2-NBDG, a fluorescently-labeled deoxyglucose analog, as a probe for the detection of glucose taken up by cultured cells. Apigenin, a flavonoid that has been reported to be an inhibitor of glucose transport, is included as a control.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty