Would you like to log in again?
Oxygen Consumption Rate Assay Kit, Cayman Chemical
Supplier: Cayman Chemical
600800-96
75817-070EA 559.57 USD InStock
75817-070
Oxygen Consumption Rate Assay Kit, Cayman Chemical
Assays Cellular Assays
For the measurement of oxygen consumption rate in living cells
  • Measure OCR without the need for an oxygen electrode
  • Includes antimycin A, an inhibitor of oxygen consumption, as a control
  • Includes glucose oxidase as a reference for oxygen depletion
The oxygen consumption rate (OCR) of cells is an important indicator of normal cellular function. It is used as a parameter to study mitochondrial function as well as a marker of factors triggering the switch from healthy oxidative phosphorylation to aerobic glycolysis in cancer cells. Oxygen consumption is traditionally measured by a cumbersome oxygen electrode, a specialized piece of equipment that typically yields low sample throughput. A Phosphorescent Oxygen Probe has proven to be useful in measuring oxygen consumption rates in whole cells. Cayman’s cell-based Oxygen Consumption Rate Assay Kit utilizes this newly developed phosphorescent oxygen probe to measure oxygen consumption rate in living cells. Antimycin A, an inhibitor of the mitochondrial electron transport chain, is included to be used as a positive control. Glucose oxidase is also included in the kit to be used as a reference for oxygen depletion. The kit is easy to use and can be easily adapted to high throughput screening for compounds which modulate oxygen consumption rate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty