Would you like to log in again?
NETosis Assay Kit, Cayman Chemical Company
Supplier: Cayman Chemical
601010-1
75817-086EA 524.89 USD InStock
75817-086
NETosis Assay Kit, Cayman Chemical Company
Assays Cellular Assays
For the measurement of NET-derived neutrophil elastase.
  • Induce and detect NET formation in vitro
  • Elastase-based readout
  • Non-dsDNA readout eliminates false positives from DNA
  • Adaptable to multiple species
Cayman's NETosis Assay Kit provides a simple and fast method for studying the process of NETosis ex vivo. Notably, Cayman's NETosis Assay Kit does not depend upon the DNA component of neutrophil extracellular traps (NETs), as DNA release can occur independently of NETosis. In this kit, primary neutrophils are stimulated to release NETs with either PMA or a calcium ionophore (both included). Unbound neutrophil elastase is washed away following NET generation. Following digest of NET DNA by S7 nuclease, the supernatant containing neutrophil elastase is added to a substrate, which is selectively cleaved by elastase to yield a 4-nitroaniline product that adsorbs light at 405 nm. Enough reagents are provided to test 24 sample conditions for NET production, with analysis in duplicate.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty