Would you like to log in again?
LDH Cytotoxicity Assay Kit, Cayman Chemical
Supplier: Cayman Chemical
601170-96 601170-480
75817-104EA 394.48 USD InStock
75817-104 75817-106
LDH Cytotoxicity Assay Kit, Cayman Chemical
Assays Cellular Assays
For measurement of LDH activity as an indicator of cell death.
  • A colorimetric, non-radioactive assay for assessing drug-induced cytotoxicity​
  • LDH release from the cytosol into the medium catalyzes a color-forming reaction
  • Detect lysis of 10,000-200,000 cells per well
  • Alternative to 51Cr-release assays
Necrosis is accompanied by mitochondrial swelling and increased plasma membrane permeability, while apoptosis involves an articulated breakdown of the cell into membrane-bound apoptotic bodies. Lactate dehydrogenase (LDH) is a soluble cytosolic enzyme that is released into the culture medium following loss of membrane integrity resulting from cell- or compound-induced cytotoxicity. LDH activity, therefore, can be used as an indicator of cell membrane integrity and serve as a general means to assess cell viability by measuring plasma membrane permeability. Cayman's LDH Cytotoxicity Assay Kit measures cell death in response to chemical compounds or environmental factors using a coupled two-step reaction. In the first step, LDH catalyzes the reduction of NAD+ to NADH and H+ by oxidation of lactate to pyruvate. In the second step of the reaction, diaphorase uses the newly-formed NADH and H+ to catalyze the reduction of a tetrazolium salt (INT) to highly-colored formazan which absorbs strongly at 490-520 nm.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty