Would you like to log in again?
Free Fatty Acid Fluorometric Assay Kit, Cayman Chemical Company
Supplier: Cayman Chemical
700310-96
75817-130EA 565.06 USD InStock
75817-130
Free Fatty Acid Fluorometric Assay Kit, Cayman Chemical Company
Assays Enzyme Assays
A sensitive tool for measuring free fatty acids in plasma, serum, and urine.
  • Measure FFAs in plasma, serum, and urine
  • Assay 20 samples in duplicate
  • Measure free fatty acids from 25 to 250 µM
  • Plate-based fluorometric measurement (ex 530 - 540 nm, em 585 - 595 nm)
The measurement of FFA can be useful in determining metabolic status. Cayman’s Free Fatty Acid (FFA) Fluorometric Assay provides a simple, reproducible, and sensitive tool for measuring free fatty acids in plasma, serum, and urine. The FFA Fluorometric Assay utilizes a coupled enzymatic reaction that results in generation of the highly fluorescent product resorufin.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty