Would you like to log in again?
Thiol Detection Assay Kit, Cayman Chemical
Supplier: Cayman Chemical
700340-96 700340-480
75817-134EA 1127.69 USD InStock
75817-134 75817-136
Thiol Detection Assay Kit, Cayman Chemical
Assays Cellular Assays
A simple, reproducible, and sensitive tool for assaying free thiol content in samples.
  • Measure free thiol content in plasma, serum, urine, cell lysates, and tissue homogenates
  • Assay 40 samples in duplicate
  • Plate-based fluorometric measurement (ex 380 – 390 nm, em 510 – 520 nm)
  • Includes both a glutathione and cysteine standard
The detection and measurement of free thiols (i.e., free cysteine, glutathione, and cysteine residues on proteins) is one of the essential tasks for investigating biological processes and events in many biological systems. Cayman’s Thiol Detection provides a simple, reproducible, and sensitive tool for assaying free thiol content in samples (i.e., plasma, serum, tissue homogenates, cell lysates, and urine). The assay utilizes a proprietary fluorometric detector that reacts with thiol groups to emit a strong fluorescent signal which can be detected using excitation wavelengths between 380 – 390 nm and emission wavelengths between 510 – 520 nm.
Delivery information: Kit includes both a glutathione and cysteine standard.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty