Would you like to log in again?
Ascorbate Assay Kit, Cayman Chemical
Supplier: Cayman Chemical
700420-96
75817-138EA 478.66 USD InStock
75817-138
Ascorbate Assay Kit, Cayman Chemical
Assays Enzyme Assays
A kit for measuring ascorbate.
  • Quantify ascorbate from plasma, serum, urine, and fruit juices
  • Assay 40 samples in duplicate
  • Plate-based fluorometric measurement (ex 340 – 350 nm, em 420 – 430 nm)
Ascorbate (L-Ascorbic acid or Vitamin C) is a six-carbon lactone that is synthesized from glucose in the liver of most mammalian species, but not by humans who must obtain ascorbate through dietary consumption. In humans, ascorbate serves as an antioxidant and acts as an electron donor for various enzymes. Cayman’s Ascorbate Assay provides a reproducible, sensitive fluorescence-based tool for quantifying ascorbate from plasma, serum, urine, and fruit juices.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty