Would you like to log in again?
L-Lactate Assay Kit, Cayman Chemical
Supplier: Cayman Chemical
700510-96
75817-146EA 544.11 USD InStock
75817-146
L-Lactate Assay Kit, Cayman Chemical
Assays Protein Assays
A fluorescence-based method for detecting L-lactate in biological samples such as serum, plasma, blood, urine, and saliva.
  • Measure L-lactate in biological samples such as serum, plasma, whole blood, urine, saliva, and cell culture
  • Assay 40 samples in duplicate
  • Plate-based fluorometric measurement (ex 530 – 540 nm, em 585 – 595 nm)
L(+)-Lactate is the major stereoisomer of lactate formed in human intermediary metabolism. The lactate to pyruvate ratio reflects the redox state of the cell and describes the balance between NAD+ and NADH, which is dependent on the interconversion of lactate and pyruvate via lactate dehydrogenase (LDH). Monitoring lactate levels is, therefore, a good way to evaluate the balance between tissue oxygen demand and utilization and is useful when studying cellular and animal physiology. Cayman’s L-Lactate Assay provides a fluorescence-based method for detecting L-lactate in biological samples such as serum, plasma, blood, urine, and saliva. It can also be utilized to determine intracellular and extracellular lactate concentrations in cell culture samples. In the assay, lactate dehydrogenase catalyzes the oxidation of lactate to pyruvate, along with the concomitant reduction of NAD+ to NADH. NADH reacts with the fluorescent substrate to yield a highly fluorescent product. The fluorescent product is analyzed with an excitation wavelength of 530 – 540 nm and an emission wavelength of 585 – 595 nm.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty