Would you like to log in again?
MitoCheck® Complex I Activity Assay Kit, Cayman Chemical
Supplier: Cayman Chemical
700930-96
75817-172EA 602.7 USD InStock
75817-172
MitoCheck® Complex I Activity Assay Kit, Cayman Chemical
Assays ELISAs
A complete system for quantifying complex I inhibition.
  • Measure complex I activity in isolated mitochondria
  • Useful for screening compounds for complex I inhibition
  • Suitable for high-throughput screening
  • Plate-based colorimetric measurement (340 nm)
  • MitoCheck® Mitochondrial (Tissue) Isolation Kit is available to aid in mitochondrial tissue isolation
Complex I (NADH oxidase/Co-enzyme Q reductase) is one of the major sites of electron entry into the mitochondrial electron transport chain (ETC). Complex I catalyzes the 2 electron oxidation of NADH followed by the reduction of ubiquinone (Q) to form ubiquinol (QH2), and ultimately the reduction of the terminal electron acceptor, O2. During the passage of electrons from NADH to Q, the translocation of four protons (H+) from the mitochondrial matrix to the intermembrane space occurs, contributing to the chemiosmotic proton gradient, which is required for oxidative phosphorylation. Cayman's MitoCheck® Complex I Activity Assay allows for the activity of complex I to be determined without the need to isolate mitochondria or pre-incubate with antibodies. The rate of NADH oxidation is measured by a decrease in absorbance at 340 nm and is proportional to the activity of complex I.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty