Would you like to log in again?
COX Colorimetric Inhibitor Screening Assay Kit, Cayman Chemical Company
Supplier: Cayman Chemical
701050-192
75817-192EA 1127.5 USD InStock
75817-192
COX Colorimetric Inhibitor Screening Assay Kit, Cayman Chemical Company
Assays ELISAs
A colorimetric assay for the screening of isozyme-specific inhibitors.
  • Screen for inhibitors of ovine COX-1 or human COX-2
  • Measure the peroxidase component of the COX
  • Assay 41 samples for COX-1 and 41 samples for COX-2 in duplicate
  • Plate-based colorimetric measurement (590 nm)
Cayman's COX Colorimetric Inhibitor Screening Assay measures the peroxidase component of cyclooxygenases. The peroxidase activity is assayed colorimetrically by monitoring the appearance of oxidized N,N,N',N'-tetramethyl-p-phenylenediamine (TMPD) at 590 nm. Inhibition of COX activity, measured by TMPD oxidation, by a variety of selective and nonselective inhibitors, shows changed potencies similar to those observed with other in vitro methods. The COX Colorimetric Inhibitor Screening assay includes both ovine COX-1 and human recombinant COX-2 enzymes in order to screen isozyme-specific inhibitors. Cayman's COX Colorimetric Assay is a time saving tool for screening vast numbers of inhibitors.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty