Would you like to log in again?
Nitrate/Nitrite Colorimetric Assay Kit (LDH method), Cayman Chemical Company
Supplier: Cayman Chemical
760871-96
75817-222EA 317.92 USD InStock
75817-222
Nitrate/Nitrite Colorimetric Assay Kit (LDH method), Cayman Chemical Company
Assays Enzyme Assays
A complete assay kit for measurement of NO2/NO3.
  • Designed to measure NOS activity in vitro
  • Can also be used to measure nitrate and nitrite in urine, plasma, serum, and tissue homogenates
  • Assay 41 samples in duplicate
  • Plate-based colorimetric measurement (530 – 550 nm)
NADPH is an essential cofactor for the function of the NOS enzyme. Unfortunately, NADPH interferes with the chemistry of the Griess reagents, which are the most commonly used reagents for nitrite detection. This kit uses Lactate Dehydrogenase (LDH) to oxidize the excess NADPH used in a NOS-catalyzed reaction, thereby making the assay particularly well suited to measurements of NOS activity in vitro. High throughput screening applications using recombinant NOS preparations can be quickly and accurately assayed with the LDH method kit.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty