Would you like to log in again?
cPLA2 Assay Kit, Cayman Chemical Company
Supplier: Cayman Chemical
765021-96
75817-228EA 730.99 USD InStock
75817-228
cPLA2 Assay Kit, Cayman Chemical Company
Assays Enzyme Assays
For the measurement of cPLA2 activity in purified preparations, cell cultures, or tissue homogenates.
  • Measure cPLA2 activity in cell lysates and tissue homogenates that are known to only contain cPLA2, or in purified enzyme preparations
  • Assay 46 samples in duplicate
  • Measure cPLA2 activity down to 0.0035 U/ml
  • Plate-based colorimetric measurement (405-420 nm)
Phospholipase A2 catalyzes the hydrolysis of fatty acids at the sn-2 position of glycerophospholipids, yielding a free fatty acid and a lysophospholipid as products
Arachidonoyl Thio-PC is a substrate for cPLA2 by virtue of the presence of arachidonic acid at the sn-2 position of the glycerophospholipid. Hydrolysis of the arachidonoyl thioester bond at the sn-2 position by PLA2 releases a free thiol which can be detected by DTNB (5,5'-dithio-bis-(2-nitrobenzoic acid)). This assay can be used to determine the activity of cPLA2 in purified preparations, cell cultures, or tissue homogenates that are known to contain only cPLA2. Use of this assay with preparations containing more than one type of PLA2 will result in the measurement of total PLA2 activity rather than cPLA2 alone. Isozyme-specific cPLA2 activity can be measured by excluding sPLA2 or inhibiting iPLA2 activities in the assay. Each kit contains cPLA2 assay buffer, DTNB/EGTA, Arachidonoyl Thio-PC (substrate), bee venom PLA2 (control), bromoenol lactone solution, a 96 well plate, and complete instructions.
The release of arachidonic acid from membrane phospholipids by these enzymes is believed to be the key step in the biosynthesis of eicosanoids. There are primarily three different kinds of phospholipase A2. They are secretory (sPLA2), calcium-dependent cytosolic (cPLA2), and calcium-independent cytosolic (iPLA2) phospholipase A2. Of these three different types of enzymes, only the cPLA2 exhibits specificity towards arachidonic acid whereas all others can hydrolyze any fatty acid at the sn-2 position.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty