Would you like to log in again?
Nitrate/Nitrite Colorimetric Assay Kit, Cayman Chemical Company
Supplier: Cayman Chemical
780001-192
75817-230EA 312.79 USD InStock
75817-230
Nitrate/Nitrite Colorimetric Assay Kit, Cayman Chemical Company
Assays Cellular Assays
For the measurement of total nitrate/nitrite concentrations.
  • Measure nitrate, nitrite, and/or total nitrate/nitrites in plasma/serum, urine, tissue culture media, and tissue homogenates
  • Assay 40 samples in duplicate/96-well plate
  • Assay Range: 5-35 µM
  • Plate-based colorimetric measurement (540-550 nm)
The Cayman Chemical Nitrate/Nitrite Assay Kit provides an accurate and convenient method for measurement of total nitrate/nitrite concentration in a simple two-step process
This kit can be used to measure nitrate and nitrite in plasma/serum, urine, tissue culture media, and tissue homogenates. However, it cannot be used to analyze nitrate and nitrite from an in vitro assay of nitric oxide synthase (NOS) in which excess NADPH has been added. For these assays a second method (LDH method) is utilized and is available from Cayman Chemical in a 96 well plate format.
The first step is the conversion of nitrate to nitrite utilizing nitrate reductase. The second step is the addition of the Griess Reagents which convert nitrite into a deep purple azo compound. Photometric measurement of the absorbance due to this azo chromophore accurately determines nitrite concentration.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty